Learn More
CBX3 Mouse anti-Human, CoraLite 488, Clone: 1A5A3, Proteintech
Product Code. 16800806 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
16800806 100 μL 100µL
1 options
Product Code. 16800806
Brand: Proteintech CL48866446100UL
Mouse Monoclonal Antibody
The CBX3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CBX3. This antibody recognizes human, mouse antigen. The CBX3 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
Antigen CBX3
Applications Immunofluorescence, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 1A5A3
Concentration 2000 μg/mL
Conjugate CoraLite 488
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 11335
Target Species Human, Mouse
Isotype IgG2a