Learn More
Abnova™ CBFB (Human) Recombinant Protein
Human CBFB full-length ORF ( AAH18509, 1 a.a. - 182 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000865-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MPRVVPDQRSKFENEEFFRKLSRECEIKYTGFGDRPHEERQARLQNACRDGRSEIAFVATGTNLSLQFFPASWQGEQRQTPSREYVDLEREAGKVYLKAPMILNGVCVIWKGWIDLQRLDGMGCLEFDEERAQQEDALAQQAFEEARRRTREFEDRDRSHREEMEVRVSQLLAVTGKKTTRP
Specifications
AAH18509 | |
core-binding factor, beta subunit | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CBFB | |
GST |
865 | |
Wheat germ expression system | |
25 μg | |
PEBP2B | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |