missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Caveolin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585274
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Dog (100%), Rhesus Monkey (100%), Bovine (100%). Positive Control: Rat muscle, mouse muscle. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Caveolin-3 (CAV3) acts as a scaffolding protein within the caveolar membrane. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-3 may also regulate voltage-gated potassium channels. It also plays a role in the sarcolemma repair mechanism of both skeletal muscle and cardiomyocytes that permits rapid resealing of membranes disrupted by mechanical stress. Caveolin-3 also mediates the recruitment of CAVIN2 and CAVIN3 proteins to the caveolae. Mutations in the gene can result in HyperCKmia, rippling muscle disease 2, cardiomyopathy, long QT syndrome 9, sudden infant death syndrome, and myopathy.
Specifications
| Caveolin 3 | |
| Polyclonal | |
| Unconjugated | |
| Cav3 | |
| AI385751; CAV; Cav3; Cav-3; caveolin 3; caveolin-3; LGMD1C; LQT9; M-cav; M-caveolin; MGC126100; MGC126101; MGC126129; structural protein; VIP21; VIP-21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12391, 29161, 859 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P51637, P51638, P56539 | |
| Cav3 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Caveolin 3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction