missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caveolin-1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 6C2B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66067-1-IG-20UL
This item is not returnable.
View return policy
Description
Caveolin-1 or CAV1 is a scaffolding proteins within caveolar membranes. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-1 is also involved in the co-stimulatory signal essential for T-cell receptor (TCR) mediated T-cell activation. It can bind with DPP4 and induces T-cell proliferation and NF-kappa-B activation in a T-cell receptor/CD3-dependent manner. Mutations affecting the CAV1 gene can result in congenital generalized lipodystrophy 3, pulmonary hypertension primary 3, or partial lipodystrophy/congenital cataracts, and neurodegeneration syndrome.Specifications
| Caveolin-1 | |
| Monoclonal | |
| 0.74 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P41350, P49817, Q03135, Q6RVA9 | |
| Mouse | |
| Protein G | |
| RUO | |
| 12389, 25404, 404693, 857 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 6C2B2 | |
| Unconjugated | |
| CAV1 | |
| CAV1 | |
| Caveolin-1 Fusion Protein Ag8049 | |
| 20 μL | |
| Primary | |
| Human, Pig, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction