missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CAV1 (Human) Recombinant Protein
Human CAV1 full-length ORF ( AAH09685, 1 a.a. - 178 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000857-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSGGKYVDSEGHLYTVPIREQGNIYKPNNKAMADELSEKQVYDAHTKEIDLVNRDPKHLNDDVVKIDFEDVIAEPEGTHSFDGIWKASFTTFTVTKYWFYRLLSALFGIPMALIWGIYFAILSFLHIWAVVPCIKSFLIEIQCISRVYSIYVHTVCDPLFEAVGKIFSNVRINLQKEI
Specifications
AAH09685 | |
caveolin 1, caveolae protein, 22kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAV1 | |
GST |
857 | |
Wheat germ expression system | |
10 μg | |
BSCL3, CAV, CGL3, MSTP085, VIP21 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |