Learn More
Cav 2.2 blocker 1, MedChemExpress
Product Code. 16487647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487647 5 mg 5mg
16461147 50 mg 50mg
16445676 100 mg 100mg
16434898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16487647 Supplier Medchem Express Supplier No. HY119373
MedChemExpress Cav 2.2 blocker 1 (compound 9) is a N-type calcium channel (Cav 2.2) blocker for the treatment of pain, with an IC50 of 1 nM.
Cav 2.2 blocker 1 (compound 9) is a N-type calcium channel (Cav 2.2) blocker for the treatment of pain, with an IC50 of 1 nM.
Specifications
Chemical Name or Material Cav 2.2 blocker 1
CAS 1567335-29-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H29ClN2O2
Quantity 5 mg
Solubility Information DMSO : 20.83 mg/mL (49.02 mM; Need ultrasonic)
SMILES COC(C=CC=C1)=C1N2C(C3=CC=C(Cl)C=C3)=CC(C4CC(C)(C)OC(C)(C)C4)=N2
Molecular Weight (g/mol) 424.96
Formula Weight 424.96
Percent Purity 99.3%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation