missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cathepsin K Monoclonal Antibody (C1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543914
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a lysosomal cysteine proteinase involved in bone remodeling and resorption. This protein, which is a member of the peptidase C1 protein family, is predominantly expressed in osteoclasts. However, the encoded protein is also expressed in a significant fraction of human breast cancers, where it could contribute to tumor invasiveness. Mutations in this gene are the cause of pycnodysostosis, an autosomal recessive disease characterized by osteosclerosis and short stature. This gene may be subject to RNA editing.
Specifications
| Cathepsin K | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O35186, P43235, P43236, P55097, Q3ZKN1, Q5E968, Q9GLE3 | |
| CTSK | |
| Ala115-Met329 (Accession P43235), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Bovine, Sheep, Pig, Horse | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C1 | |
| Unconjugated | |
| CTSK | |
| AI323530; Cat K; cathepsin K; Cathepsin O; cathepsin O1; cathepsin O2; Cathepsin X; catK; CTS02; CTSK; CTSO; CTSO1; CTSO2; MGC23107; minisatellite 10q detected by probe MMS10; MMS10-Q; Ms10q; PKND; PYCD | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100009334, 100054991, 101108972, 13038, 1513, 29175, 397569, 513038, 608843 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction