Description
- For the removal of organic components from the exhaust air.
- Organic components are catalytically oxidized at approximately 600°C, broken into carbon dioxide and water vapor.
- Irritating odors are largely eliminated.
- CE Marked
Specifications
Specifications
| For Use With (Equipment) | Nabertherm Burnout Furnaces |
| For Use With (Application) | Removal of organic components from the exhaust air |