missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Catalyst Platinum-Cured Silicone Formulation Pump Tubing
Available in standard 15m Lengths.
Brand: Fisherbrand™
Additional Details : Weight : 1.24000kg
Description
Available in standard 15m Lengths.
- Includes platinum-cured silicone, two TPEs and long-life PVC tubing material
- Available in both thin and thick wall configurations
Specifications
Catalyst pump tubing | |
8 mm | |
1.6 mm |