missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Catalyst Platinum-Cured Silicone Formulation Pump Tubing

Available in standard 15m Lengths.

Brand:  Fisherbrand™

Additional Details : Weight : 1.33000kg

 View more versions of this product

Product Code. 15501325

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Available in standard 15m Lengths.

    • Includes platinum-cured silicone, two TPEs and long-life PVC tubing material
    • Available in both thin and thick wall configurations
Specifications

Specifications

6.4 mm
2.4 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers