missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CAT1 Polyclonal antibody specifically detects CAT1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CAT1 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | amino acid transporter, cationic 1, ATRC1ERR, CAT-1System Y+ basic amino acid transporter, Ecotropic retroviral leukemia receptor homolog, ecotropic retroviral receptor, Ecotropic retrovirus receptor homolog, HCAT1, high affinity cationic amino acid transporter 1, REC1LCAT1, solute carrier family 7 (cationic amino acid transporter, y+ system), member 1, Solute carrier family 7 member 1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 530-629 of human CAT1 (NP_003036.1).,, Sequence:, TSLIAVLIITFCIVTVLGREALTKGALWAVFLLAGSALLCAVVTGVIWRQPESKTKLSFKVPFLPVLPILSIFVNVYLMMQLDQGTWVRFAVWMLIGFIIYFGYGLWHSEEASLDADQARTPDGNLDQCK |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?