Learn More
Brady™ Casters For Use With SPC Sorbent Center
Product Code. 10105884
Change view
Click to view available options
:
Daimeter: 7 in.
Product Code.
10105884 Daimeter: 7 in.
1 options
Product Code. 10105884
Brand: Brady™ 813900
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Add wheels to your SPC sorbent center to make it mobile (for use with single or double unit).
  • Set of 5 casters
  • Ø 7 cm
  • For use with SPC sorbent center - single or double unit
Specifications
Type SPC Kits
Packaging Quantity 5 Pk.
Weight (Metric) 2722 g