Learn More
Epredia™ Cassette II Slotted Tissue Cassettes in Tube Packs
Product Code. 16394056 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
Product Code. Color unitSize
16394056 Pink Pack of 750
16314056 Beige Pack of 750
16324056 Blue Pack of 750
16334056 Green Pack of 750
16344056 Gray Pack of 750
16354056 Lavender Pack of 750
16364056 Light Green Pack of 750
16374056 Lilac Pack of 750
16384056 Orange Pack of 750
16304066 Tan Pack of 750
16314066 White Pack of 750
16324066 Yellow Pack of 750
12 options
Product Code. 16394056
Brand: Epredia™ B851729PK
Slotted Cassettes are ideal for routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a front edge lid hinge, and loose-packed lids that are available in 12 color varieties.
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slots
Closure Included Yes
Quantity 750/Cs.
Closure Type Separate Lid, Front Hinge
PrintMate Compatible Yes
Color Pink
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Cassette II Biopsy Cassette