Learn More
Epredia™ Cassette II Slotted Tissue Cassettes
Product Code. 16394136 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16394136 Light Green Pack of 1000
16344136 Beige Pack of 1000
16354136 Blue Pack of 1000
16364136 Green Pack of 1000
16384136 Lavender Pack of 1000
16304146 Lilac Pack of 1000
16314146 Orange Pack of 1000
16324146 Pink Pack of 1000
16334146 Tan Pack of 1000
16344146 White Pack of 1000
16354146 Yellow Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 16394136 Supplier Epredia™ Supplier No. B851739LG
  • ONLINE EXCLUSIVE PRICE
    2360.00 NOK
    1859.77 NOK / Pack of 1000
    Save 500.23 NOK
    21% Off
 Restricted Item
In Stock
Add to basket
Add to basket
For routine tissue samples, these loose-packed cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slots
Closure Included Yes
Quantity 1000/Cs.
Closure Type Separate Lid, Front Hinge
Color Light Green
Product Type Slotted Cassette