Learn More
Epredia™ Cassette II Biopsy Tissue Cassettes in Tube Packs
Product Code. 16304076 Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
Product Code. Color unitSize
16304076 Pink Pack of 750
16244322 Blue Pack of 750
16344066 Green Pack of 750
16354066 Gray Pack of 750
16364066 Lavender Pack of 750
16374066 Light Green Pack of 750
16384066 Lilac Pack of 750
16394066 Orange Pack of 750
16314076 Tan Pack of 750
16324076 White Pack of 750
16334076 Yellow Pack of 750
11 options
Product Code. 16304076
Brand: Epredia™ B851730PK
Biopsy Cassettes are ideal for small tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a front edge lid hinge, and loose-packed lids that are available in 12 color varieties.
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Small Squares
Closure Included Yes
Quantity 750/Cs.
Closure Type Separate Lid, Front Hinge
PrintMate Compatible Yes
Color Pink
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Cassette II Biopsy Cassette