Learn More
Epredia™ Cassette II Biopsy Tissue Cassettes
Product Code. 16324126 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Product Code. Color unitSize
16324126 Beige Pack of 1000
16334126 Blue Pack of 1000
16344126 Green Pack of 1000
16354126 Gray Pack of 1000
16364126 Lavender Pack of 1000
16374126 Light Green Pack of 1000
16384126 Lilac Pack of 1000
16394126 Orange Pack of 1000
16304136 Pink Pack of 1000
16314136 Tan Pack of 1000
16324136 White Pack of 1000
16334136 Yellow Pack of 1000
12 options
Product Code. 16324126
Brand: Epredia™ B851738BE
For small tissue samples, these cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Small Squares
Closure Included Yes
Packaging Type Loose Packed (Lids)
Quantity 1000/Cs.
Closure Type Separate Lid, Front Hinge
PrintMate Compatible No
Color Beige
Product Type Cassette II with Color-Matched Lids