missing translation for 'onlineSavingsMsg'
Learn More

CASP3 Monoclonal Antibody (OTI3A12), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA813275

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17548094

  • 4065.15 NOK / 100µL
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CASP3 Monoclonal antibody specifically detects CASP3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot
Specifications

Specifications

CASP3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P42574, P55213, P70677
Mouse
Protein A/G
RUO
12367, 25402, 836
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
OTI3A12
Unconjugated
CASP3
A830040C14Rik; AC-3; Apopain; Apopain precursor; Casp3; CASP-3; caspase 3; Caspase 3 apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis related cysteine protease; Caspase 3, apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis-related cysteine peptidase; caspase 3, apoptosis-related cysteine protease; Caspase3; caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; Caspase-3-like protein; CC3; Cpp32; CPP-32; CPP32B; CPP32beta; cysteine protease CPP32; cysteine protease; CPP32; apopain; ICE3; IRP; LICE; mLdy; PARP cleavage protease; procaspase3; protein Yama; SCA-1; SREBP cleavage activity 1; Yama; yama protein
Human recombinant protein fragment corresponding to amino acids 29-219 of human CASP3 (NP_116786) produced in E.coli.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers