Learn More
Carvedilol-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Carvedilol-methyl-d3 |
| CAS | 1020719-25-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 2H3 H23 N2 O4 |
| Quantity | 10 mg |
| Synonym | 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-[2-(methoxy-d3)phenoxy]ethyl]amino]- (ACI), 1-(9H-Carbazol-4-yloxy)-3-[[2-[2-(methoxy-d3)phenoxy]ethyl]amino]-2-propanol (ACI), 1-(9H-Carbazol-4-yloxy)-3-[2-[2-(trideuteriomethoxy)phenoxy]ethylamino]propan-2-ol, Carvedilol-d3, Carvedilol-D3 (methoxy-D3) |
| InChI Formula | InChI=1S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27H,13-16H2,1H3/i1D3 |
| SMILES | [2H]C([2H])([2H])Oc1ccccc1OCCNCC(O)COc2cccc3[nH]c4ccccc4c23 |
| IUPAC Name | 1-(9H-carbazol-4-yloxy)-3-[2-[2-(trideuteriomethoxy)phenoxy]ethylamino]propan-2-ol |
| Molecular Weight (g/mol) | 409.49 |
| Show More |