Learn More
Cartap Hydrochloride, TRC
Product Code. 30134326
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134326 250 mg 250mg
30040766 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30134326 Supplier Toronto Research Chemicals Supplier No. TRCC184453250MG
 Restricted Item
Estimated Shipment: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15263-52-2
Molecular Formula C7 H15 N3 O2 S2 . Cl H
Molecular Weight (g/mol) 273.8
Synonym Carbamic acid, thio-, S,S'-[2-(dimethylamino)trimethylene] ester, monohydrochloride (8CI), Carbamothioic acid, S,S'-[2-(dimethylamino)-1,3-propanediyl] ester, monohydrochloride (9CI), 1,3-Propanedithiol, 2-(dimethylamino)-, dicarbamate (ester), monohydrochloride (8CI), 1,3-Bis(carbamoylthio)-2-(N,N-dimethylamino)propane hydrochloride, 2-(Dimethylamino)-1,3-propanedithiol dicarbamate hydrochloride, Caldan, Cartap hydrochloride, NTD 2, Padan, Padan 4G, Padan SG, Suntap, Vegetox
IUPAC Name S-[3-carbamoylsulfanyl-2-(dimethylamino)propyl] carbamothioate;hydrochloride
SMILES Cl.CN(C)C(CSC(=O)N)CSC(=O)N
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C7H15N3O2S2.ClH/c1-10(2)5(3-13-6(8)11)4-14-7(9)12;/h5H,3-4H2,1-2H3,(H2,8,11)(H2,9,12);1H
Formula Weight 273.0372
Percent Purity >95%
Chemical Name or Material Cartap hydrochloride
Grade HPLC