missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CARS (Human) Recombinant Protein
Human CARS full-length ORF ( AAH02880, 1 a.a. - 748 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000833-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MADSSGQQGKGRRVQPQWSPPAGTQPCRLHLYNSLTRNKEVFIPQDGKKVTWYCCGPTVYDASHMGHARSYISFDILRRVLKDYFKFDVFYCMNITDIDDKIIKRARQNHLFEQYREKRPEAAQLLEDVQAALKPFSVKLNETTDPDKKQMLERIQHAVQLATEPLEKAVQSRLTGEEVNSCVEVLLEEAKDLLSDWLDSTLGCDVTDNSIFSKLPKFWEGDFHRDMEALNVLPPDVLTRVSEYVPEIVNFVQKIVDNGYGYVSNGSVYFDTAKFASSEKHSYGKLVPEAVGDQKALQEGEGDLSISADRLSEKRSPNDFALWKASKPGEPSWPCPWGKGRPGWHIECSAMAGTLLGASMDIHGGGFDLRFPHHDNELAQSEAYFENDCWVRYFLHTGHLTIAGCKMSKSLKNFITIKDALKKHSARQLRLAFLMHSWKDTLDYSSNTMESALQYEKFLNEFFLNVKDILRAPVDITGQFEKWGEEEAELNKNFYDKKTAIHKALCDNVDTRTVMEEMRALVSQCNLYMAARKAVRKRPNQALLENIALYLTHMLKIFGAVEEDSSLGFPVGGPGTSLSLEATVMPYLQVLSEFREGVRKIAREQKVPEILQLSDALRDNILPELGVRFEDHEGLPTVVKLVDRNTLLKEREEKRRVEEEKRKKKEEAARRKQEQEAAKLAKMKIPPSEMFLSETDKYSKFDENGLPTHDMEGKELSKGQAKKLKKLFEAQEKLYKEYLQMAQNGSFQ
Specifications
AAH02880 | |
cysteinyl-tRNA synthetase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CARS | |
GST |
833 | |
Wheat germ expression system | |
25 μg | |
CARS1, CYSRS, MGC:11246 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |