Learn More
Thermo Scientific™ Carrying Kit Set for Eutech™ Cond 6+ and TDS 6+ meters
Product Code. 12613135
Change view
Click to view available options
For Use With (Equipment):
Eutech Cond 6+ and TDS 6+ Meters
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12613135 Eutech Cond 6+ and TDS 6+ Meters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12613135 Supplier Thermo Scientific™ Supplier No. 01X266902
  • ONLINE EXCLUSIVE PRICE
    905.00 NOK
    832.95 NOK / Pack of 1
    Save 72.05 NOK
    7% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Eutech™ Carrying Kit Cases are thoughtfully designed to hold your Eutech™ meter and electrodes.
Eutech™ Carrying Kit Cases are thoughtfully designed to hold Eutech™ meters and electrodes.The Economy Carrying Kit Set is a plastic carrying case for the Eutech™ Cond 6+ and TDS 6+ meters and probes, calibration solutions, batteries, and manuals.
The Carrying Kit set is comprised of:
  • 1413 μS/CM KCL Calibration Solution
  • 12.88 mS/cm KCl Calibration Solution
  • 3000 ppm 442 TDS standard solutions
  • Deionised rinse water (60 ml)
  • TRUSTED_SUSTAINABILITY
    Specifications
    Description Carrying Kit Set for Eutech™ Cond 6+ and TDS 6+ meters
    Type Accessory
    Includes 1413 μS/CM KCL Calibration Solution, 12.88 mS/cm KCl Calibration Solution, 3000 ppm 442 TDS standard solutions, deionised rinse water (60 ml)
    For Use With (Equipment) Eutech Cond 6+ and TDS 6+ Meters