Learn More
Hettich™ Carriers for Hettich Rotors 1624 and 1726
Description
- Rotor Adapter for Hettich Drum Rotors
- For use with Rotina, Rotofix and Universal Centrifuges
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
- ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
- ROTINA Centrifuges are ideally equipped to deliver separation technology to satisfy the highest standards
Specifications
Specifications
| Type | Carrier for Rotor |
| Capacity | 5 x 9 mL |
| Max. Tube Diameter | 14 mm |
| Max. Tube Length | 100 mm |
| For Use With (Equipment) | Hettich Rotors 1624 and 1726 |
| Vessels per Adapter | 5 |