Learn More
Hettich Lab Technology™ Carrier for Swing-out Rotors
Click to view available options
For Use With (Equipment):
Hentich Swing-out Rotors
Unit Size:
Each
Description
- Designed for use with Hettich Swing-out Rotors
- Holds 15 to 17 mm dia. tubes
Specifications
Specifications
| For Use With (Equipment) | Hentich Swing-out Rotors |
| Bore Size (Metric) | 17.5 mm |
| Tube Size | 15 to 17 mm |