Learn More
Hettich™ Carrier for Hettich Rotor no. 4274 and 4474
Click to view available options
Capacity:
4 x 50 to 100 mL Tubes
Unit Size:
Each
Description
- Carrier for Hettich Rotors
- Holds ASTM tubes and 50mL Babcock flask
- For use with Rotors no. 4274 and 4474
- Compatible with ROTANTA 460/460 R/460 RC/460 RF, ROTOFIX 46/46 H, ROTIXA 500 RS Centrifuges
Specifications
Specifications
| Type | Carrier for Rotor |
| Tube Type | ASTM and Babcock |
| Capacity | 4 x 50 to 100 mL Tubes |
| Max. Tube Diameter | 58 mm |
| Max. Tube Length | 200 mm |
| For Use With (Equipment) | Hettich Rotor |
| Vessels per Adapter | 4 |
| No. of Places | 4 |