Learn More
Hettich™ Carrier for Hettich Rotor no. 1628
Product Code. 10247822
Click to view available options
Capacity:
7.5 to 15 mL Tubes
Unit Size:
Each
Product Code. 10247822
Brand: Hettich™ 1621
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Carrier for Hettich Rotor no. 1628
  • For use with ROTOFIX 32 A and Universal 320/320 R Centrifuge
Specifications
Type Carrier for Rotor
Tube Type Round Bottom, Conical, Blood/Urine, Cryo
Capacity 7.5 to 15 mL Tubes
Max. Tube Diameter 17 mm
Max. Tube Length 100 mm
For Use With (Equipment) Hettich Rotor
Vessels per Adapter 1
No. of Places 1