Learn More
Hettich™ Carrier for Hettich Rotor no. 1611
Product Code. 10052683
Change view
Click to view available options
Capacity:
1 x 4 to 10 mL Tube
1 x 7.5 to 15 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
10052683 1 x 7.5 to 15 mL Each
12023884 1 x 4 to 10 mL Tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10052683 Supplier Hettich™ Supplier No. 1644
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Carrier for Hettich Rotor no. 1611
  • For use with ROTOFIX 32 A and Universal 320/320 R Centrifuge, 8-place
Specifications
Type Carrier for Rotor
Tube Type Blood/Urine, Round Bottom
Capacity 1 x 7.5 to 15 mL
Max. Tube Diameter 17 mm
Max. Tube Length 100 mm
For Use With (Equipment) Hettich Rotor
Vessels per Adapter 1