Learn More
Hettich™ Carrier for Hettich Rotor no. 1016 and 1137
Click to view available options
Capacity:
50 mL
Unit Size:
Each
Description
- Carrier for Hettich Rotor no. 1016 and 1137
- For use with EBA 280/280 S and MIKRO 220/220 R
Specifications
Specifications
| Type | Carrier for Rotor |
| Tube Type | Round Bottom |
| Capacity | 50 mL |
| Max. Tube Diameter | 29 mm |
| Max. Tube Length | 107 mm |
| For Use With (Equipment) | Hettich Rotor |
| Vessels per Adapter | 1 |
| No. of Places | 1 |