Learn More
Motic™ Widefield Eyepiece WF15X/12mm
Product Code. 11838183
Click to view available options
Magnification Power:
15X
Unit Size:
Each
Product Code. 11838183
Brand: Motic™ 53100143
Only null left
Add to basket
Add to basket
Change the standard 10X eyepiece of your microscope. Motic™ Widefield eyepiece WF15X/12mm will increase magnification by 15X.
  • Increases standard 10X magnification
  • Field Number: 12mm
  • 15X magnification
Specifications
Type Widefield Eyepiece
Field of View 12 mm
Magnification Power 15X
Quantity 1 Ea.