Learn More
Motic™ CCIS™ LWD Plan Achromatic Objective
Description
- For use with AE31 Inverted Microscope
- Long Working Distance
- Achromatic Plan Objective
- Flat field
- Eliminates color aberration
- Magnification and Numerical Apertures available:
- 20X/0.3 (WD=4.7mm)
- 40X/0.5 (WD=3.0mm)
Specifications
Specifications
| Type | Plan |
| Focus Distance | 0.5 mm, WD=3.0 mm |
| For Use With (Equipment) | AE inverted microscopes |
| Magnification Power | 40X |
| Quantity | 1 Piece |