Learn More
Motic™ Micrometer Eyepiece Reticle
Product Code. 11732298
Change view
Click to view available options
Type:
Reticle Eyepiece Micrometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11732298 Reticle Eyepiece Micrometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11732298 Supplier Motic™ Supplier No. 1101001401841
  • ONLINE EXCLUSIVE PRICE
    562.00 NOK
    487.08 NOK / Pack of 1
    Save 74.92 NOK
    13% Off
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
Use with 10X eyepiece for BA Series Microscope. Motic™ Micrometer Eyepiece Reticle includes 100 divisions in 10mm
  • 100 divisions in 10mm
  • 25mm dia.
  • For use with BA series microscope 10X eyepiece
Specifications
Type Reticle Eyepiece Micrometer
Description 100 divisions in 10mm
Diameter (Metric) 25 mm
For Use With (Equipment) BA Series Microscope
Quantity 1 Piece