Learn More
Abnova™ CARHSP1 (Human) Recombinant Protein
Product Code. 16127502
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16127502
Brand: Abnova™ H00023589P01.10ug
Human CARHSP1 full-length ORF ( NP_001035941.1, 1 a.a. - 147 a.a.) recombinant protein with GST-tag at N-terminal.
  • Sequence: MSSEPPPPPQPPTHQASVGLLDTPRSRERSPSPLRGNVVPSPLPTRRTRTFSATVRASQGPVYKGVCKCFCRSKGHGFITPADGGPDIFLHISDVEGEYVPVEGDEVTYKMCSIPPKNEKLQAVEVVITHLAPGTKHETWSGHVISS
Specifications
Accession Number NP_001035941.1
Gene ID (Entrez) 23589
Name calcium regulated heat stable protein 1, 24kDa
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CRHSP-24, CSDC1, MGC111446
Gene Symbol CARHSP1
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer