Learn More
- Home
- Products
- Boxes
- Cryogenic and Freezer Boxes
- ratiolab™ Cardboard Grid Insert for Cryo Boxes
ratiolab™ Cardboard Grid Insert for Cryo Boxes
Product Code. 10003063
Change view
Click to view available options
Array:
10 x 10
12 x 12
13 x 13
14 x 14
16 x 16
5 x 5
6 x 6
7 x 7
9 x 9
Height (Metric):
25 mm
40 mm
65 mm
Length (Metric):
133 mm
136 mm
Width (Metric):
133 mm
136 mm
Unit Size:
Pack of 10
14 product options available for selection
| Product Code. | Array | Height (Metric) | Length (Metric) | Width (Metric) | Unit Size |
|---|---|---|---|---|---|
| 10003063 | 14 x 14 | 40 mm | 133 mm | 133 mm | Pack of 10 |
| 10162513 | 5 x 5 | 25 mm | 133 mm | 133 mm | Pack of 10 |
| 15592065 | 6 x 6 | 25 mm | 133 mm | 133 mm | Pack of 10 |
| 10262913 | 6 x 6 | 65 mm | 136 mm | 136 mm | Pack of 10 |
| 17303931 | 7 x 7 | 25 mm | 133 mm | 133 mm | Pack of 10 |
| 10843211 | 9 x 9 | 65 mm | 133 mm | 133 mm | Pack of 10 |
| 15617375 | 10 x 10 | 65 mm | 133 mm | 133 mm | Pack of 10 |
| 10130273 | 12 x 12 | 65 mm | 136 mm | 136 mm | Pack of 10 |
| 10190123 | 12 x 12 | 40 mm | 136 mm | 136 mm | Pack of 10 |
| 16350816 | 13 x 13 | 40 mm | 133 mm | 133 mm | Pack of 10 |
| 10395922 | 13 x 13 | 25 mm | 136 mm | 136 mm | Pack of 10 |
| 10395702 | 14 x 14 | 65 mm | 133 mm | 133 mm | Pack of 10 |
| 13427477 | 14 x 14 | 25 mm | 136 mm | 136 mm | Pack of 10 |
| 10229044 | 16 x 16 | 25 mm | 136 mm | 136 mm | Pack of 10 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
This item is not returnable.
View return policy
Product Code.
10003063
Supplier
ratiolab™
Supplier No.
5120206
You must Sign In and have a business account to purchase this item.
Restricted Item
Estimated Shipment: 30-09-2026
Log in to see stock.
Log in to see stock.
Add to basket
Add to basket
This item is not returnable.
View return policy
Description
- For sample storage, even at ultra-low temperatures
- Temperature-resistant down to -80°C
- Water-repellent finish
- Packaging in practical folding boxes
Specifications
Specifications
| Quantity | 10 Inserts/Pk. |
| Material | Cardboard |
| Product Type | Box Divider Insert |
| Height (Metric) | 40 mm |
| For Use With (Equipment) | 7 mm Dia. Tubes |
| Width (Metric) | 133 mm |
| Length (Metric) | 133 mm |
| Array | 14 x 14 |