Learn More
- Home
- Products
- Boxes
- Cryogenic and Freezer Boxes
- Tenak™ Cardboard Cryoboxes
Tenak™ Cardboard Cryoboxes
Product Code. 15280514 Shop All TENAK Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Dimensions (L x W x H):
75 x 75 mm
100 x 100 mm
133 x 133 x 32 mm
133 x 133 x 50 mm
133 x 133 x 52 mm
133 x 133 x 75 mm
136 x 136 x 32 mm
136 x 136 x 50 mm
136 x 136 x 75 mm
133 x 133 x 100 mm
133 x 133 x 130 mm
Includes:
10 x 10 Divider
9 x 9 Divider
Dividers
Unit Size:
Each
Pack of 36
23 product options available for selection
| Product Code. | Color | Dimensions (L x W x H) | Includes | unitSize |
|---|---|---|---|---|
| 15280514 | White | 133 x 133 x 32 mm | Dividers | Pack of 36 |
| 11597294 | Blue | 133 x 133 x 50 mm | Dividers | Pack of 36 |
| 15130887 | Red | 133 x 133 x 50 mm | Dividers | Pack of 36 |
| 11521705 | White | 75 x 75 mm | Dividers | Each |
| 15280554 | White | 75 x 75 mm | Dividers | Pack of 36 |
| 15170877 | Yellow | 133 x 133 x 50 mm | Dividers | Pack of 36 |
| 15200604 | White | 136 x 136 x 75 mm | Dividers | Pack of 36 |
| 15250594 | White | 136 x 136 x 50 mm | Dividers | Pack of 36 |
| 11620443 | White | 133 x 133 x 52 mm | 10 x 10 Divider | Pack of 36 |
| 15200594 | White | 136 x 136 x 32 mm | Dividers | Pack of 36 |
| 15210604 | Blue | 133 x 133 x 75 mm | Dividers | Pack of 36 |
| 15220594 | Red | 133 x 133 x 32 mm | Dividers | Pack of 36 |
| 15220524 | White | 133 x 133 x 75 mm | Dividers | Pack of 36 |
| 15220604 | Red | 133 x 133 x 75 mm | Dividers | Pack of 36 |
| 15230604 | Green | 133 x 133 x 75 mm | Dividers | Pack of 36 |
| 15260534 | White | 133 x 133 x 100 mm | Dividers | Pack of 36 |
| 15270554 | White | 133 x 133 x 52 mm | 9 x 9 Divider | Pack of 36 |
| 11511705 | White | 133 x 133 x 52 mm | 9 x 9 Divider | Each |
| 15280534 | Red | 133 x 133 x 100 mm | Dividers | Pack of 36 |
| 11531705 | White | 100 x 100 mm | Dividers | Each |
| 15290554 | White | 100 x 100 mm | Dividers | Pack of 36 |
| 15290604 | White | 133 x 133 x 130 mm | 9 x 9 Divider | Pack of 36 |
| 15210524 | White | 133 x 133 x 50 mm | Dividers | Pack of 36 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Cardboard
- Temperature resistant down to -80°C
- Ideal for ultra-low temperature storage
Specifications
Specifications
| Color | White |
| Material | Cardboard |
| Type | Cryo Box |
| For Use With (Application) | For Cryogenic Storage |
| Includes | Dividers |
| Quantity | 36 Pack |
| Dimensions (L x W x H) | 133 x 133 x 32 mm |
| Length (Metric) | 133 mm |
| Width (Metric) | 133 mm |
| Height (Metric) | 32 mm |