Learn More
Tenak™ Origami Cardboard Cryobox
Shop All TENAK Products
Click to view available options
Height (Metric):
50 mm
52 mm
Includes:
10 x 10 Divider
9 x 9 Divider
No divider
Unit Size:
Pack of 100
Description
- Spacesaving in the laboratory
- Square measure of 134x134mm fits all storage rack systems
- Packages of 100 each
Specifications
Specifications
| Material | Cardboard |
| Shape | Square |
| Type | Cryo Box |
| Includes | 10 x 10 Divider |
| Quantity | 100 Pack |
| Width (Metric) | 134 mm |
| Height (Metric) | 50 mm |
| Weight (Metric) | 0.05 kg |