Learn More
- Home
- Products
- Boxes
- Cryogenic and Freezer Boxes
- Ratiolab™ Cardboard Cryo Boxes With Waterproof Plastic Coating
Ratiolab™ Cardboard Cryo Boxes With Waterproof Plastic Coating
Product Code. p-8013974
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Length (Metric):
133 mm
136 mm
Width (Metric):
133 mm
136 mm
Height (Metric):
32 mm
50 mm
75 mm
100 mm
130 mm
Unit Size:
Pack of 10
| Product Code. | Color | Length (Metric) | Width (Metric) | Height (Metric) | unitSize |
|---|---|---|---|---|---|
| 10212343 | Blue | 136 mm | 136 mm | 100 mm | Pack of 10 |
| 10553575 | Red | 136 mm | 136 mm | 100 mm | Pack of 10 |
| 10434363 | Green | 136 mm | 136 mm | 100 mm | Pack of 10 |
| 10632254 | White | 136 mm | 136 mm | 100 mm | Pack of 10 |
| 10232483 | Yellow | 136 mm | 136 mm | 100 mm | Pack of 10 |
| 12625785 | Blue | 133 mm | 133 mm | 130 mm | Pack of 10 |
| 10707123 | Blue | 136 mm | 136 mm | 130 mm | Pack of 10 |
| 10115974 | White | 133 mm | 133 mm | 130 mm | Pack of 10 |
| 10501085 | White | 136 mm | 136 mm | 130 mm | Pack of 10 |
| 10304633 | Yellow | 136 mm | 136 mm | 130 mm | Pack of 10 |
| 10150803 | White | 136 mm | 136 mm | 32 mm | Pack of 10 |
| 15293979 | Blue | 133 mm | 133 mm | 32 mm | Pack of 10 |
| 10799054 | Yellow | 136 mm | 136 mm | 32 mm | Pack of 10 |
| 10222673 | Blue | 136 mm | 136 mm | 50 mm | Pack of 10 |
| 10691674 | Green | 136 mm | 136 mm | 50 mm | Pack of 10 |
| 10141093 | Red | 136 mm | 136 mm | 50 mm | Pack of 10 |
| 15210568 | White | 133 mm | 133 mm | 50 mm | Pack of 10 |
| 10797453 | White | 136 mm | 136 mm | 50 mm | Pack of 10 |
| 15971641 | Yellow | 133 mm | 133 mm | 50 mm | Pack of 10 |
| 10406283 | Yellow | 136 mm | 136 mm | 50 mm | Pack of 10 |
| 10707293 | Green | 136 mm | 136 mm | 75 mm | Pack of 10 |
| 10141043 | White | 136 mm | 136 mm | 75 mm | Pack of 10 |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Ultra low temperature-resistant cardboard
- For sample storage
- Temperature-resistant down to -80°C
- Water-repellent finish
- Packaging in practical folding boxes
Specifications
Specifications
| Color | Blue |
| Material | Cardboard |
| Type | Cryo Box |
| For Use With (Application) | Tubes from dia. 6mm to dia. 28.5mm |
| Holds | Tubes (6 mm to 28.5 mm dia.) |
| Quantity | 10 Pack |
| Length (Metric) | 136 mm |
| Width (Metric) | 136 mm |
| Height (Metric) | 100 mm |