Learn More
Invitrogen™ Carboxy-DCFDA (5-(and-6)-Carboxy-2',7'-Dichlorofluorescein Diacetate), mixed isomers
Beskrivning
Carboxy-DCFDA can passively diffuse into cells and is colorless and nonfluorescent until the acetate groups are cleaved by intracellular esterases to yield the fluorescent fluorophore, 5-(and-6)-carboxy-2',7' -dichlorofluorescein.
Carboxy-DCFDA can passively diffuse into cells and is colorless and nonfluorescent until the acetate groups are cleaved by intracellular esterases to yield the fluorescent fluorophore, 5-(and-6)-carboxy-2',7' -dichlorofluorescein. With a pKa of 4.8, this dye can be used as an acidic pH sensor.
To make a concentrated stock solution from 1–10 mM, dissolve in either high quality anhydrous DMSO, DMF, or 100% ethanol. Once prepared, use the stock solutions within a short time period.
Beställningsinfo
Shipping Condition: Room temperature
Specifikationer
Specifikationer
| Content And Storage | Store in freezer (-5°C to -30°C). |
| Detection Method | Fluorescence |
| For Use With (Application) | Polar Tracer, pH Indicator |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Carboxy-DCFDA |
| Dye Type | pH Indicator |
| Quantity | 100 mg |
| Shipping Condition | Room Temperature |
| Molecular Weight (g/mol) | 529.29 |
For Research Use Only. Not for use in diagnostic procedures.
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.