Learn More
Swann-Morton™ Carbon Steel Sterile Scalpel Blades
Product Code. 11728363
Change view
Click to view available options
Size:
# 10
# 11
# 15
# 18
# 21
# 22
# 23
# 24
Unit Size:
Pack of 100
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11728363 # 10 Pack of 100
11877550 # 18 Pack of 100
11778363 # 21 Pack of 100
11738363 # 11 Pack of 100
11758363 # 15 Pack of 100
11798363 # 23 Pack of 100
11792724 # 22 Pack of 100
11708373 # 24 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 11728363 Supplier Swann-Morton™ Supplier No. 0201
  • ONLINE EXCLUSIVE PRICE
    256.00 NOK
    222.01 NOK / Pack of 100
    Save 33.99 NOK
    13% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Use for convenience in user environments: on the ward, in emergency or trauma units or even out in the field with paramedics. Swann Morton™ Carbon Steel Surgical Scalpel Blades come in single peel packs.
  • Carbon steel to BS EN 27740
  • Sterile
Specifications
Blade Material Carbon Steel
Product Type Scalpel Blade
Size # 10
For Use With (Equipment) The # 10 fits Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L
Sterility Sterile
Certifications/Compliance CE compliant 93/42/EC
Quantity 10/Pk.