Learn More
Swann-Morton™ Carbon Steel Scalpel Blades
Description
Use Swann Morton™ Carbon Steel Scalpel Blades in the operating room for precise cutting and repair.
- Carbon steel to BS EN 27740
- Handles and blades sold separately
Specifications
Specifications
| Blade Material | Carbon Steel |
| Product Type | Scalpel Blade |
| For Use With (Application) | Aesthetic Procedures such as Dermaplaning which helps to refinish the Skin's top layers through a method of Controlled Surgical Scraping |
| Size | # 14 |
| Sterility | Non-sterile |
| Quantity | 10/Pk. |