Learn More
Swann-Morton™ Carbon Steel Scalpel Blades
Product Code. 11320915
Change view
Click to view available options
Size:
# 14
# 20
# 22A
# 27
# 36
60 x 10.96 mm
Major-Z
Unit Size:
Pack of 100
Pack of 50
Product Code. Size unitSize
11320915 60 x 10.96 mm Pack of 50
11768363 # 20 Pack of 100
12474070 # 27 Pack of 100
11837383 # 14 Pack of 100
12308019 # 22A Pack of 100
11841733 # 36 Pack of 100
12922545 Major-Z Pack of 50
7 options
Product Code. 11320915
Brand: Swann-Morton™ 4211
Only null left
Add to basket
Add to basket
Use Swann Morton™ Carbon Steel Scalpel Blades in the operating room for precise cutting and repair.
  • Carbon steel to BS EN 27740
  • Handles and blades sold separately
Specifications
Blade Material Carbon Steel
Product Type SM 01
For Use With (Application) Tough Cutting Jobs
Size 60 x 10.96 mm
Length (Metric) 60 mm
For Use With (Equipment) SCA-760-V Type
Sterility Non-sterile
Thickness (Metric) 0.63 mm
Quantity 50/Pk.