Learn More
Swann-Morton™ Carbon Steel Scalpel Blades
Description
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
- Carbon steel to BS EN 27740
- Handles and blades sold separately
Specifications
Specifications
| Blade Material | Carbon Steel |
| Product Type | Scalpel Blade |
| Size | # 22 |
| Length (Metric) | 40 mm |
| For Use With (Equipment) | Larger than the # 20 and # 21 the # 22 fits Handles # 4,# 4L, # 4 Graduated and # 6B |
| Disposable | Single-use |
| Certifications/Compliance | BS EN 7740 |
| Quantity | 10/Pk. |