Learn More
Swann-Morton™ Carbon Steel Scalpel Blades
Description
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
- Carbon steel to BS EN 27740
- Handles and blades sold separately
Specifications
Specifications
| Blade Material | Carbon Steel |
| Product Type | Scalpel Blade |
| Size | # 15 |
| Length (Metric) | 15 mm |
| For Use With (Equipment) | The # 15 Blade fit Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L |
| Disposable | Single-use |
| Certifications/Compliance | BS EN 7740 |
| Quantity | 10/Pk. |