Learn More
Swann-Morton™ Carbon Steel Scalpel Blades
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
Brand:  Swann-Morton™ 0103
Additional Details : Weight : 0.08100kg
 View more versions of this product
Product Code. 11748373
  • 241.00 NOK / Pack of 100
Estimated Shipment: 27-05-2024
Description
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
  • Carbon steel to BS EN 27740
  • Handles and blades sold separately
Specifications
Specifications
Carbon Steel
# 11
The # 11 Blade fit Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L
BS EN 7740
Scalpel Blade
20 mm
Yes
10/Pk.