Learn More
Swann-Morton™ Carbon Steel Scalpel Blades
Product Code. 11702734
Change view
Click to view available options
Size:
# 10
# 10A
# 11
# 15
# 20
# 21
# 22
# 23
# 24
Length (Metric):
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
Unit Size:
Pack of 100
Product Code. Size Length (Metric) unitSize
11702734 # 23 30 mm Pack of 100
11738373 # 10 25 mm Pack of 100
12901635 # 10A 15 mm Pack of 100
11748373 # 11 20 mm Pack of 100
11762724 # 15 15 mm Pack of 100
11772724 # 20 30 mm Pack of 100
12118931 # 22 40 mm Pack of 100
11782724 # 21 35 mm Pack of 100
11712734 # 24 35 mm Pack of 100
9 options
Product Code. 11702734
Brand: Swann-Morton™ 0110
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
  • Carbon steel to BS EN 27740
  • Handles and blades sold separately
Specifications
Blade Material Carbon Steel
Product Type Scalpel Blade
Size # 23
Length (Metric) 30 mm
For Use With (Equipment) The # 23 fits Handles # 4, # 4L, # 4 Graduated and # 6B
Disposable Single-use
Certifications/Compliance BS EN 7740
Quantity 10/Pk.