missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Resulfurized, AISI 1144, G11440, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Carbon Steel, Resulfurized, AISI 1144, G11440, 100g
Specifications
Specifications
| Chemical Name or Material | Lead/Boron/Niobium/Titanium/Aluminium/Tungsten/Oxygen/Vanadium/Arsenic/Cobalt/Nitrogen/Tin/Phosphorus/Molybdenum/Nickel/Chromium/Copper/Silicon/Carbon/Manganese/Zirconium/Selenium/Magnesium/Zinc/Tantalum/Calcium/Antimony |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Lead 0.001 %/Boron 0.0012 %/Niobium 0.0016 %/Titanium 0.0016 %/Aluminium 0.0019 %/Tungsten 0.0023 %/Oxygen 0.0037 %/Vanadium 0.0037 %/Arsenic 0.0059 %/Cobalt 0.007 %/Nitrogen 0.007 %/Tin 0.0084 %/Phosphorus 0.0155 %/Molybdenum 0.0293 %/Nickel 0.085 %/Chromium 0.19 %/Copper 0.193 %/Silicon 0.21 %/Carbon 0.469 %/Manganese 1.55 %/Zirconium (<0.0040) %/Selenium (<0.006) %/Magnesium (0.0002) %/Zinc (0.0006) %/Tantalum (0.0007) %/Calcium (0.0011) %/Antimony (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction