Learn More
Swann-Morton™ Carbon Steel Postmortem Scalpel Blades
Product Code. p-8000786
Change view
Click to view available options
Size:
# PM40
# PM40 B
Product Type:
Blunted Knife
Sharp
Unit Size:
Pack of 10
Product Code. Size Product Type unitSize
12952555 # PM40 B Blunted Knife Pack of 10
11360905 # PM40 Sharp Pack of 10
2 options
Product Code. 12952555
Brand: Swann-Morton™ 2552
Only null left
Add to basket
Add to basket
Use Swann Morton™ Carbon Steel Postmortem Scalpel Blades to facilitate pathology operations.
  • Added safety feature of a 'bull nose tip' to help offset the occurrence of accidental sharp related injuries
  • Blunted
  • PM-40
Specifications
Blade Material Carbon Steel
Product Type Blunted Knife
For Use With (Application) For Postmortem procedures with the added safety feature of a 'bull nose tip' to help offset the occurrence of Accidental Sharp related Injuries
Size # PM40 B
For Use With (Equipment) The PM40 Stainless, PM40 Standard or PM40 Plated Handle