Learn More
Swann-Morton™ Carbon Steel Postmortem Scalpel Blades
Product Code. 11360905
Change view
Click to view available options
Size:
# PM40
# PM40 B
Product Type:
Blunted Knife
Sharp
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Product Type Unit Size
11360905 # PM40 Sharp Pack of 10
12952555 # PM40 B Blunted Knife Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11360905 Supplier Swann-Morton™ Supplier No. 2551
  • ONLINE EXCLUSIVE PRICE
    204.00 NOK
    165.60 NOK / Pack of 10
    Save 38.40 NOK
    18% Off
Estimated Shipment: 08-12-2026
Use Swann Morton™ Carbon Steel Postmortem Scalpel Blades to facilitate pathology operations.
  • Added safety feature of a 'bull nose tip' to help offset the occurrence of accidental sharp related injuries
  • Heavy duty carbon steel blade
  • Metal handle with screw-type fitting
  • PM-40
.
Specifications
Blade Material Carbon Steel
Product Type Sharp
Size # PM40
For Use With (Equipment) SCA-710-K Swann-Morton
Description Sharp
Quantity 10/Pk.