Learn More
Erlab™ Captair Smart Trespa™ Work Surface
Description
- Made of phenolic resin with ergonomic arm for a comfortable working position
- High chemical and mechanical resistance
- Ideal for precision weighing operations
Specifications
Specifications
| For Use With (Equipment) | Smart 392 Fume Hood |
| Includes | Spill Tray |
| Depth (Metric) | 644 mm |
| Width (Metric) | 875 mm |
| Product Type | Work Surface |
| Material | Phenolic Resin |