Learn More
Erlab™ Captair Smart Glass Work Surface with Retention Tray
Product Code. p-7160208
Change view
Click to view available options
For Use With (Equipment):
Smart 321 Fume Hood
Smart 391 Fume Hood
Smart 392 Fume Hood
Smart 481 Fume Hood
Smart 483 Fume Hood
Smart 633 Fume Hood
Smart 714 Fume Hood
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15535154 Smart 321 Fume Hood Each
15565154 Smart 391 Fume Hood Each
15595154 Smart 481 Fume Hood Each
15512904 Smart 392 Fume Hood Each
15525164 Smart 483 Fume Hood Each
15555164 Smart 633 Fume Hood Each
15575164 Smart 714 Fume Hood Each
7 options
Product Code. 15535154
Brand: Erlab™ I0160020001
Erlab™ Captair Smart Work Surface with Retention Tray is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
  • Tempered enameled glass worktop
  • Includes retention tray filtering fume hoods
  • Jointed frame
  • Ergonomic arm rest for a comfortable working position
Specifications
For Use With (Equipment) Smart 321 Fume Hood
Includes Retention Tray
Depth (Metric) 470 mm
Description Jointed Frame
Width (Metric) 700 mm
Product Type Work Surface
Material Tempered Glass