Learn More
Erlab™ Captair by Erlab™ Stainless Steel Work Surfaces
Click to view available options
For Use With (Equipment):
CaptairFlow 391 Ductless Fume Hood
CaptairFlow 483 Ductless Fume Hood
CaptairFlow 714 Ductless Fume Hood
Unit Size:
Each
Description
- High chemical and mechanical resistance
- Rounded corners to facilitate cleaning operations
- Built in spill tray
Specifications
Specifications
| Material | Stainless Steel |
| For Use With (Equipment) | CaptairFlow 714 Ductless Fume Hood |