Learn More
BRAND™ Caps for Transferpettor™
Product Code. 10430993 Shop All Brand Products
Click to view available options
Volume (Metric):
5 μL
20 μL
25 μL
Unit Size:
Pack of 100
Product Code. 10430993
Brand: BRAND™ 701906
Only null left
Add to basket
Add to basket
DE-M certified. Includes color code. Glass construction.
  • For use with Transferpettor™
  • DE-M marking
  • Glass construction
  • Includes color code
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DE-M certified
Color White
Material Glass
Volume (Metric) 25 μL
Quantity 100 Pack